missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKB zeta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56390SS
This item is not returnable.
View return policy
Description
IKB zeta Polyclonal specifically detects IKB zeta in Mouse samples. It is validated for Western Blot.
Specifications
| IKB zeta | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 64332 | |
| Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml | |
| Q9BYH8, Q9EST8 | |
| NFKBIZ | |
| Synthetic peptides corresponding to an amino acid range between 270-320 and 670-720 of mouse IkB zeta protein were used as the immunogen for this antibody. | |
| 0.025 mg | |
| Primary | |
| This antibody is reactive against Mouse. Predicted cross-reactivity based on sequence identity: Mouse (100%), Human, (100%), Rat (100%), Chimp (100%), Bovine (100%), Chicken (93%). Not yet tested in other species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction