missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKAROS Polyclonal Antibody

Rabbit Polyclonal Antibody
£550.00
Specifications
| Antigen | IKAROS |
|---|---|
| Concentration | 0.3 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17178973
![]() |
Invitrogen™
PA5111054 |
100 μL |
£550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17188973
|
Invitrogen™
PA5111055 |
100 μL |
£550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: DADEGQDMSQ VSGKESPPVS DTPDEGDEPM PIPEDLSTTS GGQQSSKSDR VVASNVKVET QSDEENGRAC EMNGEECAED LRMLDASGEK MNGSH Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 92%.
DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.Specifications
| IKAROS | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| IKZF1 | |
| 5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1 | |
| IKZF1 | |
| Primary | |
| Antigen affinity chromatography |
| 0.3 mg/mL | |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13422 | |
| 10320 | |
| Recombinant Protein corresonding to Human IKAROS. Recombinant protein control fragment (Product #RP-102656). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title