missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKAROS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111055
This item is not returnable.
View return policy
Description
Immunogen sequence: DADEGQDMSQ VSGKESPPVS DTPDEGDEPM PIPEDLSTTS GGQQSSKSDR VVASNVKVET QSDEENGRAC EMNGEECAED LRMLDASGEK MNGSH Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 92%.
DNA-binding protein IKAROS functions in the specification and the maturation of the T lymphocyte. It also interacts with a critical control element in the terminal deoxynucleotidyl transferase promoter as well as with the promoters for other genes expressed during early stages of B-cell and T-cell development.
Specifications
| IKAROS | |
| Polyclonal | |
| Unconjugated | |
| IKZF1 | |
| 5832432G11Rik; CLL-associated antigen KW-6; CVID13; DNA-binding protein Ikaros; hIk-1; hlk-1; Hs.54452; IK1; IKAROS; IKAROS family zinc finger 1; IKAROS family zinc finger 1 (Ikaros); ikaros family zinc finger protein 1; IKZF1; KAROS family zinc finger 1 (Ikaros); LYF1; LyF-1; Lymphoid transcription factor LyF-1; mKIAA4227; PPP1R92; PRO0758; protein phosphatase 1, regulatory subunit 92; RGD1562979; Zfpn1a1; zinc finger containing transcription factor; zinc finger protein, subfamily 1A, 1 (Ikaros); Znfn1a1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13422 | |
| IKZF1 | |
| Recombinant Protein corresonding to Human IKAROS. Recombinant protein control fragment (Product #RP-102657). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction