missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ikaros/IKZF1 Rabbit anti-Human, Mouse, Clone: 4S10I3, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | Ikaros/IKZF1 |
|---|---|
| Clone | 4S10I3 |
| Dilution | Western Blot 1:50 - 1:200, Chromatin Immunoprecipitation 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ChIP Assay, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18344065
|
Novus Biologicals
NBP3-16223-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18337955
|
Novus Biologicals
NBP3-16223-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ikaros/IKZF1 Monoclonal antibody specifically detects Ikaros/IKZF1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, ImmunofluorescenceSpecifications
| Ikaros/IKZF1 | |
| Western Blot 1:50 - 1:200, Chromatin Immunoprecipitation 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Ikaros/IKZF1 (Q13422). AEDLCKIGSERSLVLDRLASNVAKRKSSMPQKFLGDKGLSDTPYDSSASYEKENEMMKSHVMDQAINNAINYLGAESLRPLVQTPPGGSEVVPVISPMYQL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 4S10I3 | |
| ChIP Assay, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10320 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title