missing translation for 'onlineSavingsMsg'
Learn More

IKarisoside A, MedChemExpress

Product Code. 16411568
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411568 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411568 Supplier Medchem Express Supplier No. HYN0875
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IKarisoside A (Icarisoside-A) is a natural flavonol glycoside and has anti-inflammatory properties.

IKarisoside A (Icarisoside-A) is a natural flavonol glycoside and has anti-inflammatory properties.

Specifications

Chemical Name or Material IKarisoside A
CAS 55395-07-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H28O10
Quantity 5 mg
Synonym Icarisoside-A Baohuoside II
Solubility Information DMSO : 100 mg/mL (199.80 mM; Need ultrasonic)
SMILES OC1=CC(O)=C2C(OC(C3=CC=C(O)C=C3)=C(O[C@H](O[C@@H](C)[C@H](O)[C@H]4O)[C@@H]4O)C2=O)=C1C/C=C(C)\C
Molecular Weight (g/mol) 500.49
Formula Weight 500.49
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.