missing translation for 'onlineSavingsMsg'
Learn More

IKA™ IKAFLON™ Alnico V Stir Bars

Brand:  IKA™ 0001129000

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10411201

  • £124.00 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

27 mm
155mm
1/Ea.
Sphere
PTFE
Stir Bar
Alnico Magnet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers