missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF2/CD101 Antibody (BB27) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £402.00
Specifications
| Antigen | IGSF2/CD101 |
|---|---|
| Clone | BB27 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18098153
|
Novus Biologicals
NB100-65269-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18083414
|
Novus Biologicals
NB100-65269 |
0.1 mg |
£402.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| IGSF2/CD101 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2 | |
| CD101 | |
| IgG1 | |
| Protein G purified | |
| NB100-65269 recognizes human CD101. CD101 is a 140kD homodimeric cell surface glycoprotein expressed primarily by monocytes, granulocytes, dendritic cells and activated T lymphocytes. Studies suggest that CD101 plays a major role in the activation of T cells by skin dendritic cells. Clone BB27 has been reported to inhibit allogeneic T-cell responses (1). Removal of sodium azide is recommended prior to use in functional assays. |
| BB27 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q93033 | |
| 9398 | |
| Human thymic clone B12 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title