missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF2/CD101 Antibody (BB27) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65269-0.025MG
This item is not returnable.
View return policy
Description
IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| IGSF2/CD101 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Q93033 | |
| CD101 | |
| Human thymic clone B12 | |
| 0.025 mg | |
| Primary | |
| NB100-65269 recognizes human CD101. CD101 is a 140kD homodimeric cell surface glycoprotein expressed primarily by monocytes, granulocytes, dendritic cells and activated T lymphocytes. Studies suggest that CD101 plays a major role in the activation of T cells by skin dendritic cells. Clone BB27 has been reported to inhibit allogeneic T-cell responses (1). Removal of sodium azide is recommended prior to use in functional assays. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunohistochemistry | |
| BB27 | |
| Unconjugated | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9398 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction