missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (II/41), PE/Cy7, Novus Biologicals™
Rat Monoclonal Secondary Antibody
£257.00 - £330.00
Specifications
| Antigen | Mouse IgM |
|---|---|
| Clone | II/41 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217351
|
Novus Biologicals
NBP1-42940-0.05MG |
0.05 mg |
£257.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244041
|
Novus Biologicals
NBP1-42940-0.1MG |
0.1 mg |
£330.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.Specifications
| Mouse IgM | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| RUO | |
| Mouse | |
| Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
| IGHM | |
| IgG2a κ | |
| Affinity Purified |
| II/41 | |
| Flow Cytometry | |
| Monoclonal | |
| Rat | |
| Immunology | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
| 3507 | |
| The immunogen for this antibody was IgM. | |
| Secondary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title