missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (II/41), PE/Cy7, Novus Biologicals™
Rat Monoclonal Secondary Antibody
Brand: Novus Biologicals NBP1-42940-0.05MG
212.50 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Mouse IgM | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| II/41 | |
| PE-Cyanine7 | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
| IGHM | |
| The immunogen for this antibody was IgM. | |
| 0.05 mg | |
| Immunology | |
| 3507 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction