missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Goat anti-Mouse, CoraLite594, Secondary Antibody, Proteintech
Goat anti-Mouse Affinipure Secondary Antibody
£40.00 - £84.00
Specifications
| Antigen | IgG(H+L) |
|---|---|
| Applications | Immunofluorescence, Flow Cytometry |
| Conjugate | CoraLite 594 |
| Form | Liquid |
| Host Species | Goat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17650397
|
Proteintech
SA00013-3-200UL |
200 μL |
£40.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17660397
|
Proteintech
SA00013-3-500UL |
500 μL |
£84.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IgG(H+L) | |
| CoraLite 594 | |
| Goat | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| Store at -20°C. Stable for one year after shipment. |
| Immunofluorescence, Flow Cytometry | |
| Liquid | |
| Mouse | |
| Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title