Learn More
IgG(H+L) Goat anti-Mouse, CoraLite594, Secondary Antibody, Proteintech
Goat anti-Mouse Affinipure Secondary Antibody
Brand:  Proteintech SA00013-3-200UL
 View more versions of this product
Product Code. 17650397
  • £40.00 / 200µL
Specifications
IgG(H+L)
CoraLite 594
Goat
200 μL
Mouse
Liquid
Immunofluorescence, Flow Cytometry
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Protein A was purified by chromatography from Staphylococcus aureus
Secondary
Store at -20°C. Stable for one year after shipment.