missing translation for 'onlineSavingsMsg'
Learn More

IgG Antibody (E20-V), Novus Biologicals™

Rabbit Monoclonal Antibody

£377.00 - £507.00

Specifications

Antigen IgG
Clone E20-V
Dilution Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18658507
View Documents
Novus Biologicals
NBP3-08196-200ul
200 μg
£507.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18651667
View Documents
Novus Biologicals
NBP3-08196-100ul
100 μg
£377.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

IgG
Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300
Monoclonal
Purified
RUO
Human
Immunoglobulin G, ImmunoglobulinG
Peptide derived from internal domain of human IgG-1 chain C region. Antibody recognizes the epitope betweenVal167 - Val185.
Primary
Store at 4°C.
E20-V
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Adaptive Immunity, Epitope Tags, Immunology
20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA
3500
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.