missing translation for 'onlineSavingsMsg'
Learn More

IgG Antibody (E20-V), Novus Biologicals™

Product Code. 18658507 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18658507 200 μg 200µL
18651667 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18658507 Supplier Novus Biologicals Supplier No. NBP308196200ul
  • ONLINE EXCLUSIVE PRICE
    £507.00
    £478.80 / 200µL
    Save £28.20
    5% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E20-V
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100-1:300, Immunohistochemistry-Paraffin 1:100-1:300
Formulation 20 mM Tris-HCl, pH 8.0, 20 mg/ml BSA
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Peptide derived from internal domain of human IgG-1 chain C region. Antibody recognizes the epitope betweenVal167 - Val185.
Purification Method Immunogen affinity purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.