missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (RM0096-11B16) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£153.00 - £426.00
Specifications
| Antigen | IGFBP-3 |
|---|---|
| Clone | RM0096-11B16 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18269852
|
Novus Biologicals
NBP1-22452-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18272650
|
Novus Biologicals
NBP1-22452 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.Specifications
| IGFBP-3 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Apoptosis, Prostate Cancer | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| IGFBP3 | |
| IgG2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RM0096-11B16 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 3486 | |
| Mouse recombinant protein of IGFBP-3 | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title