missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (RM0096-11B16) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22452-0.025MG
This item is not returnable.
View return policy
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Mouse samples. It is validated for Western Blot.
Specifications
| IGFBP-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000 | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of IGFBP-3. | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0096-11B16 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| IGFBP3 | |
| Mouse recombinant protein of IGFBP-3 | |
| 0.025 mg | |
| Apoptosis, Prostate Cancer | |
| 3486 | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction