missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFALS/ALS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | IGFALS |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
IGFALS/ALS Polyclonal specifically detects IGFALS/ALS in Human samples. It is validated for Western Blot.Specifications
| IGFALS | |
| Polyclonal | |
| Rabbit | |
| P35858 | |
| 3483 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ALSinsulin-like growth factor binding protein complex acid labile chain, insulin-like growth factor binding protein, acid labile subunit, insulin-like growth factor-binding protein complex acid labile subunit | |
| IGFALS | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title