missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68981
This item is not returnable.
View return policy
Description
IGF2BP3 Polyclonal specifically detects IGF2BP3 in Mouse samples. It is validated for Western Blot.
Specifications
| IGF2BP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CT98, hKOC, IGF II mRNA binding protein 3, IGF2 mRNA-binding protein 3, IGF-II mRNA-binding protein 3, IMP-3KH domain-containing protein overexpressed in cancer, IMP3VICKZ family member 3, insulin-like growth factor 2 mRNA binding protein 3, insulin-like growth factor 2 mRNA-binding protein 3, KH domain containing protein overexpressed in cancer, KOC1DKFZp686F1078, VICKZ3cancer/testis antigen 98 | |
| Rabbit | |
| 63 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Guinea pig: 100%; Equine: 92%; Rabbit: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9CPN8 | |
| IGF2BP3 | |
| Synthetic peptides corresponding to Igf2bp3 (insulin-like growth factor 2 mRNA binding protein 3) The peptide sequence was selected from the middle region of Igf2bp3 (NP_076159). Peptide sequence QQNPSPQLRGRRGPGQRGSSRQASPGSVSKQKPCDLPLRLLVPTQFVGAI. | |
| Affinity purified | |
| RUO | |
| 10643 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction