missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I R/IGF1R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£170.00 - £391.00
Specifications
| Antigen | IGF-I R |
|---|---|
| Concentration | 1.05 mg/mL |
| Dilution | Western Blot 1 ug/ml, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067595
|
Novus Biologicals
NBP1-77679SS |
0.025 mL |
£170.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745943
|
Novus Biologicals
NBP1-77679 |
0.1 mL |
£391.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IGF-I R | |
| Western Blot 1 ug/ml, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| IGF1R | |
| IgG | |
| Affinity Purified |
| 1.05 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| P08069 | |
| 3480 | |
| A synthetic peptide made to a C-terminal portion of the human IGF1 Receptor protein (between residues 1300-1367) [UniProt P08069]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title