missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I R/IGF1R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77679SS
This item is not returnable.
View return policy
Description
IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IGF-I R | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.05 mg/mL | |
| Western Blot 1 ug/ml, Simple Western 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
| P08069 | |
| IGF1R | |
| A synthetic peptide made to a C-terminal portion of the human IGF1 Receptor protein (between residues 1300-1367) [UniProt P08069]. | |
| 0.025 mL | |
| Apoptosis, Cancer, Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3480 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction