Learn More
Invitrogen™ Mouse IgD Monoclonal Antibody (11-26c (11-26)), Alexa Fluor™ 660, eBioscience™

Rat Monoclonal Antibody
£107.00 - £224.00
Specifications
| Antigen | Mouse IgD |
|---|---|
| Clone | 11-26c (11-26) |
| Concentration | 0.2 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30535496
|
Invitrogen™
606-5993-80 |
25 μg |
£107.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30534471
![]() |
Invitrogen™
606-5993-82 |
100 μg |
£224.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor 660 emits at 690 nm and is intended for use on spectral cytometers where it may be multiplexed with APC, Alexa Fluor 647 and Alexa Fluor 700. Excitation: 662 nm; Emission: 690 nm; Laser: Red Laser.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
| Mouse IgD | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Alexa Fluor 660 | |
| Rat | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P01881 | |
| 380797 | |
| Primary | |
| Affinity chromatography |
| 11-26c (11-26) | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Mouse | |
| Ighd | |
| IgD; Igh-5; IGHD; immunoglobulin heavy constant delta | |
| Ighd | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.