missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgD Monoclonal Antibody (11-26c (11-26)), Alexa Fluor™ 660, eBioscience™
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ 606-5993-82

 View more versions of this product

Product Code. 30534471

  • £224.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Alexa Fluor 660 emits at 690 nm and is intended for use on spectral cytometers where it may be multiplexed with APC, Alexa Fluor 647 and Alexa Fluor 700. Excitation: 662 nm; Emission: 690 nm; Laser: Red Laser.

This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgD
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P01881
Ighd
Affinity chromatography
RUO
380797
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
11-26c (11-26)
Alexa Fluor 660
Ighd
IgD; Igh-5; IGHD; immunoglobulin heavy constant delta
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.