missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | IFT88 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
IFT88 Polyclonal specifically detects IFT88 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IFT88 | |
| Polyclonal | |
| Rabbit | |
| NP_006522 | |
| 8100 | |
| Synthetic peptide directed towards the middle region of human IFT88The immunogen for this antibody is IFT88. Peptide sequence EIREQRIKSGRDGSGGSRGKREGSASGDSGQNYSASSKGERLSARLRALP. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10 | |
| IFT88 | |
| IgG | |
| 91 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title