missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79538
This item is not returnable.
View return policy
Description
IFT88 Polyclonal specifically detects IFT88 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IFT88 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10 | |
| Rabbit | |
| 91 kDa | |
| 100 μL | |
| Primary | |
| Predicted Homology Based On Immunogen Sequence: Rat: 86%; Rabbit: 86%; Pig: 85%; Horse: 85%; Mouse: 85%; Guinea pig: 85%. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_006522 | |
| IFT88 | |
| Synthetic peptide directed towards the middle region of human IFT88The immunogen for this antibody is IFT88. Peptide sequence EIREQRIKSGRDGSGGSRGKREGSASGDSGQNYSASSKGERLSARLRALP. | |
| Affinity purified | |
| RUO | |
| 8100 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction