Learn More
IFN-γ Rat anti-Mouse, FITC, Clone: XMG1.2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554411
Additional Details : Weight : 0.01000kg
Description
The XMG1.2 antibody reacts with mouse interferon-γ (IFN-γ) protein. IFN-γ is a pleiotropic cytokine, of approximately 15-17kDa, involved in the regulation of the immune response. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN-γ production is associated with the Th-1 differentiation. This is a neutralizing antibody
Host Species: Rat
Clone: XMG1.2
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse IFN-γ
Intracellular Staining
Specifications
IFN-γ | |
Monoclonal | |
0.5mg/mL | |
IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
XMG1.2 | |
FITC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse IFN-γ | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 κ |
For Research Use Only.