missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-γ Rat anti-Mouse, FITC, Clone: XMG1.2, BD
Rat Monoclonal Antibody
£171.00 - £380.00
Specifications
Antigen | IFN-γ |
---|---|
Clone | XMG1.2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15821569
|
BD Biosciences
562019 |
25 μg |
£171.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15847678
|
BD Biosciences
554411 |
0.1 mg |
£380.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The XMG1.2 monoclonal antibody specifically binds to mouse interferon- γ(IFN- γ) protein. IFN- γis a pleiotropic cytokine, of approximately 15-17 kDa, involved in the regulation of inflammatory and immune responses. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN- γproduction is associated with the Th1 cell differentiation. The purified form of this antibody has been reported to be a neutralizing antibody.
Specifications
IFN-γ | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG1 κ | |
Affinity Purified | |
IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon |
XMG1.2 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.