missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN gamma Monoclonal Antibody (XMG1.2), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Product Code. 15350840
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15350840 100 μg 100µg
15340840 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15350840 Supplier Invitrogen™ Supplier No. 17731182

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rat Monoclonal Antibody

Description: The XMG1.2 antibody reacts with mouse interferon-gamma. The XMG1.2 antibody is a neutralizing antibody. Mouse IFN-gamma is a 20 kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN-gamma, in synergy with other cytokines such as TNF-alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN-gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN-gamma. Glycosylation of IFN-gamma does not affect its biological activity. Applications Reported: This XMG1.2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: The APC conjugated XMG1.2 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN gamma
Applications Flow Cytometry
Classification Monoclonal
Clone XMG1.2
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IFNG
Gene Accession No. P01580
Gene Alias BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Gene Symbols IFNG
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15978
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.