missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN beta Recombinant Rabbit Monoclonal Antibody (23GB6230), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | IFN beta |
|---|---|
| Clone | 23GB6230 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087624
![]() |
Invitrogen™
MA553002 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089904
|
Invitrogen™
MA553003 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN beta Recombinant Monoclonal antibody specifically detects IFN beta in Human, Mouse samples. It is validated for Western BlotSpecifications
| IFN beta | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P01574, P01575 | |
| 15977, 3456 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6230 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1 | |
| A synthesized peptide derived from human Interferon beta (50-110AA). | |
| Primary | |
| -20°C | |
| Ifnb1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title