missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN beta Recombinant Rabbit Monoclonal Antibody (23GB6230), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553003
This item is not returnable.
View return policy
Description
Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
Specifications
| IFN beta | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P01574, P01575 | |
| Ifnb1 | |
| A synthesized peptide derived from human Interferon beta (50-110AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 23GB6230 | |
| Unconjugated | |
| Ifnb1 | |
| Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15977, 3456 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction