Learn More
Invitrogen™ IFN beta Human ELISA Kit
Measure levels of human interferon beta (IFNb) in human cell and tissue culture media supernatant samples using 96-well plates.
£862.00 - £1767.58
Specifications
Description | Human IFN beta ELISA Kit |
---|---|
Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
Specificity | No significant cross-reaction with human IFN-a or IFN-g; mouse or rat IFN-ß |
Measurement | Absorbance at 450nm |
Precision | Intra-assay CV ≤ 10%; inter-assay CV ≤ 10% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11885804
|
Invitrogen™
414101 |
96 Tests |
£862.00
1 set |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10403615
|
Invitrogen™
414102 |
5 Plate Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Assay IFN-b (human interferon beta) levels in human cell and tissue culture media (supernatants) with theseInvitrgen™ Human IFN beta ELISA Kits.
These human IFN beta ELISA kits use pre-titered, matched pairs of coating and detection antibodies to accurately measure interferon beta (IFN-β) in samples at 100μL per well. These IFN-β kits are validated for use with human cell and tissue culture media. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.Target: Human interferon beta (human IFN-β, IFN-beta)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well plate
Measurement: Absorbance at 450nm
Assay Range: 25 to 2000pg/mL
Sensitivity: Undetermined
Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Types: Cell culture supernatant, tissue culture media
Total Assay Time: 3.25 hours
Sample Size: 100μL
Specificity: No significant cross-reaction with human IFN-α or IFN-γ; mouse or rat IFN-β
Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, substrates, and stop solution.
Specifications
Human IFN beta ELISA Kit | |
No significant cross-reaction with human IFN-a or IFN-g; mouse or rat IFN-ß | |
Intra-assay CV ≤ 10%; inter-assay CV ≤ 10% |
Colorimetric ELISA (TMB substrate) with Plates | |
Absorbance at 450nm |