missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IFN beta ELISA Kit

Product Code. 11885804
Change view
Click to view available options
Quantity:
5 Plate Kit
96 Tests
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11885804 96 Tests 1 set
10403615 5 Plate Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 11885804 Supplier Invitrogen™ Supplier No. 414101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

ELISA

The Human Interferon beta (Hu IFNβ) ELISA quantitates Hu IFNβ in human cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IFNβ. Principle of the method The Human IFNβ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01574
Assay Range 50 to 4000 pg/mL
Assay Sensitivity 50 pg/mL
Description Human IFN beta ELISA Kit
Conjugate HRP
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA
Sample Type Supernatant
Specificity No significant cross-reaction with human IFN-a or IFN-g; mouse or rat IFN-ß
Sufficient For 96 microplate wells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3456
Gene Symbol IFB,IFF,IFN-beta,IFNB
Interassay CV <8%
Intraassay CV <8
Kit Contents Pre-coated 96 well Microplate, Detection Antibody Concentrate, Anti-IgG-HRP Concentrate, Concentrate Diluent, Standard, Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Measurement Absorbance at 450 nm
Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IFN beta
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 15 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.