missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha B2/IFN-alpha 8 Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | IFN-alpha B2/IFN-alpha 8 |
|---|---|
| Clone | 3 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231346
|
Novus Biologicals
NBP2-89440-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257397
|
Novus Biologicals
NBP2-89440-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha B2/IFN-alpha 8 Monoclonal specifically detects IFN-alpha B2/IFN-alpha 8 in Human samples. It is validated for ELISA.Specifications
| IFN-alpha B2/IFN-alpha 8 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3445 | |
| IgG | |
| Protein A purified |
| 3 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B′, Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B | |
| Recombinant Human IFN-alpha B2/IFN-alpha 8 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title