missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha B2/IFN-alpha 8 Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89440-100UL
This item is not returnable.
View return policy
Description
IFN-alpha B2/IFN-alpha 8 Monoclonal specifically detects IFN-alpha B2/IFN-alpha 8 in Human samples. It is validated for ELISA.
Specifications
| IFN-alpha B2/IFN-alpha 8 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3445 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 3 | |
| ELISA 1:5000-1:10000 | |
| IFN-alpha-8, IFN-alphaB, interferon alpha type 201, interferon alpha-8, interferon alpha-B′, Interferon alpha-B, Interferon alpha-B2, interferon, alpha 8, LeIF B | |
| Recombinant Human IFN-alpha B2/IFN-alpha 8 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction