missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£260.00
Specifications
| Antigen | IFN-alpha 2 |
|---|---|
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IFN-alpha 2 Polyclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Western Blot, ELISA.Specifications
| IFN-alpha 2 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P01563 | |
| 3440 | |
| The immunogen for this antibody was a sequence from 25a.a. to 186a.a. from Interferon alpha 2b(a 188a.a. long protein) | |
| Affinity Purified | |
| Interferon alpha 2b |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Apoptosis, Cytokine Research | |
| PBS with 0.02% Sodium Azide | |
| alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
| IFNA2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title