missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75556-0.025mg
This item is not returnable.
View return policy
Description
IFN-alpha 2 Polyclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| IFN-alpha 2 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| P01563 | |
| IFNA2 | |
| The immunogen for this antibody was a sequence from 25a.a. to 186a.a. from Interferon alpha 2b(a 188a.a. long protein) | |
| 0.025 mg | |
| Apoptosis, Cytokine Research | |
| 3440 | |
| Human |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Interferon alpha 2b | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction