missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Mouse anti-Human, Clone: 1948-02-01 00:00:00, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | IFN-alpha 1 |
|---|---|
| Clone | 1948-02-01 00:00:00 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Frozen |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Description
IFN-alpha 1 Monoclonal specifically detects IFN-alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| IFN-alpha 1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| Purified recombinant human IFN-alpha 1 | |
| Primary | |
| Store at 4°C. |
| 1948-02-01 00:00:00 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3439 | |
| IgG1 κ | |
| Protein A or G purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel