missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Mouse anti-Human, Clone: 1948-02-01 00:00:00, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07232-100ug
This item is not returnable.
View return policy
Description
IFN-alpha 1 Monoclonal specifically detects IFN-alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| IFN-alpha 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 1948-02-01 00:00:00 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Purified recombinant human IFN-alpha 1 | |
| 100 μg | |
| Cancer, Immunology, Innate Immunity | |
| 3439 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction