missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFITM2/3 Mouse anti-Human, Clone: 2E8D12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | IFITM2/3 |
|---|---|
| Clone | 2E8D12 |
| Concentration | 0.85 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16837635
|
Proteintech
66081-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827635
|
Proteintech
66081-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody recognizes both IFITM2 and IFITM3.
IFN-induced antiviral protein that mediates cellular innate immunity to at least three major human pathogens, namely influenza A H1N1 virus, West Nile virus (WNV), and dengue virus (WNV), by inhibiting the early step(s) of replication.Specifications
| IFITM2/3 | |
| 0.85 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 1 8U, IFITM3, IP15 | |
| IFITM2, IFITM3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2E8D12 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q01628, Q01629 | |
| 10410, 10581 | |
| IFITM2/3 Fusion Protein Ag17863 | |
| Primary | |
| -20°C | |
| IFITM3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title