missing translation for 'onlineSavingsMsg'
Learn More

IFITM2/3 Mouse anti-Human, Clone: 2E8D12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66081-1-IG-20UL

 View more versions of this product

Product Code. 16837635

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody recognizes both IFITM2 and IFITM3.

IFN-induced antiviral protein that mediates cellular innate immunity to at least three major human pathogens, namely influenza A H1N1 virus, West Nile virus (WNV), and dengue virus (WNV), by inhibiting the early step(s) of replication.
Specifications

Specifications

IFITM2/3
Monoclonal
0.85 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01628, Q01629
IFITM2, IFITM3
IFITM2/3 Fusion Protein Ag17863
20 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry
2E8D12
Unconjugated
IFITM3
1 8U, IFITM3, IP15
Mouse
Protein G
RUO
10410, 10581
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.