missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | IFI27L2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
IFI27L2 Polyclonal specifically detects IFI27L2 in Human samples. It is validated for Western Blot.Specifications
| IFI27L2 | |
| Polyclonal | |
| Rabbit | |
| Q9H2X8 | |
| 83982 | |
| Synthetic peptides corresponding to FAM14A(family with sequence similarity 14, member A) The peptide sequence was selected from the middle region of FAM14A. Peptide sequence SVGSVLGACLGNSPSSSLPAEPEAKEDEARENVPQGEPPKPPLKSEKHEE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FAM14A, family with sequence similarity 14, member A, interferon alpha-inducible protein 27-like protein 2, interferon, alpha-inducible protein 27-like 2, Interferon-stimulated gene 12b protein, ISG12(b), ISG12(b) protein, ISG12B, pIFI27-like protein, Protein TLH29, TLH29MGC44913 | |
| IFI27L2 | |
| IgG | |
| 12 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title