missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62510
This item is not returnable.
View return policy
Description
IFI27L2 Polyclonal specifically detects IFI27L2 in Human samples. It is validated for Western Blot.
Specifications
| IFI27L2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FAM14A, family with sequence similarity 14, member A, interferon alpha-inducible protein 27-like protein 2, interferon, alpha-inducible protein 27-like 2, Interferon-stimulated gene 12b protein, ISG12(b), ISG12(b) protein, ISG12B, pIFI27-like protein, Protein TLH29, TLH29MGC44913 | |
| Rabbit | |
| 12 kDa | |
| 100 μL | |
| Primary | |
| Bovine: 77%. | |
| Human, Bovine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H2X8 | |
| IFI27L2 | |
| Synthetic peptides corresponding to FAM14A(family with sequence similarity 14, member A) The peptide sequence was selected from the middle region of FAM14A. Peptide sequence SVGSVLGACLGNSPSSSLPAEPEAKEDEARENVPQGEPPKPPLKSEKHEE. | |
| Affinity purified | |
| RUO | |
| 83982 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction