Learn More
Invitrogen™ IDO Chimeric Recombinant Mouse Monoclonal Antibody (7H8L17), Invitrogen™
Mouse Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | IDO Chimeric |
|---|---|
| Clone | 7H8L17 |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30131680
|
Invitrogen™
702743M20UG |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30131964
|
Invitrogen™
702743M |
100 μL |
£360.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody has Rabbit Variable heavy and light chains, and Mouse constant domains. It recognizes human IDO protein. This chimeric antibody is recognized by anti-mouse secondary antibodies and has no cross reactivity to anti-rabbit secondary antibodies. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
IDO Chimeric Recombinant Monoclonal antibody specifically detects IDO Chimeric in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| IDO Chimeric | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
| Protein corresponding to human IDO (aa1-aa403). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ido1 |
| 7H8L17 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P14902 | |
| 3620 | |
| IgG | |
| Protein A/G | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.