Learn More
Invitrogen™ IDO Chimeric Recombinant Mouse Monoclonal Antibody (7H8L17), Invitrogen™
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702743M20UG
Description
This antibody has Rabbit Variable heavy and light chains, and Mouse constant domains. It recognizes human IDO protein. This chimeric antibody is recognized by anti-mouse secondary antibodies and has no cross reactivity to anti-rabbit secondary antibodies. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.
Specifications
| IDO Chimeric | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P14902 | |
| Ido1 | |
| Protein corresponding to human IDO (aa1-aa403). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 7H8L17 | |
| Unconjugated | |
| Ido1 | |
| EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.