missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IDE Recombinant Rabbit Monoclonal Antibody (23GB6210), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | IDE |
|---|---|
| Clone | 23GB6210 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087452
![]() |
Invitrogen™
MA553950 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089739
|
Invitrogen™
MA553951 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IDE Recombinant Monoclonal antibody specifically detects IDE in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| IDE | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P14735, P35559, Q9JHR7 | |
| 15925, 25700, 3416 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6210 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
| A synthesized peptide derived from human IDE (31-86AA). | |
| Primary | |
| -20°C | |
| IDE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title