missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IDE Recombinant Rabbit Monoclonal Antibody (23GB6210), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553951
This item is not returnable.
View return policy
Description
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
Specifications
| IDE | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IDE | |
| 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15925, 25700, 3416 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6210 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14735, P35559, Q9JHR7 | |
| IDE | |
| A synthesized peptide derived from human IDE (31-86AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction