missing translation for 'onlineSavingsMsg'
Learn More

ID4 Antibody (PCRP-ID4-1A1), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen ID4
Clone PCRP-ID4-1A1
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 μg/mL, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate
Applications Western Blot, ELISA, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18670365
View Documents
Novus Biologicals
NBP3-20624-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18607304
View Documents
Novus Biologicals
NBP3-20624-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications

Specifications

ID4
0.2 mg/ml
Western Blot, ELISA, Immunoprecipitation
Unconjugated
Mouse
Human
bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein
Recombinant fragment (around aa 62-112) of human ID4 protein
Primary
Store at 4&drg;C. Do not freeze.
PCRP-ID4-1A1
Western Blot 1-2 μg/mL, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate
Monoclonal
Purified
RUO
10mM PBS with 0.05% BSA
3400
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.