missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID4 Antibody (PCRP-ID4-1A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20624-100ug
This item is not returnable.
View return policy
Description
ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| ID4 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate | |
| bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
| Recombinant fragment (around aa 62-112) of human ID4 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| PCRP-ID4-1A1 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3400 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction