missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55014
This item is not returnable.
View return policy
Description
ICT Polyclonal specifically detects ICT in Human samples. It is validated for Western Blot.
Specifications
| ICT | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Digestion substraction 1, DS-1DS1, EC 3.1.1.29, immature colon carcinoma transcript 1, Immature colon carcinoma transcript 1 protein, peptidyl-tRNA hydrolase ICT1, mitochondrial | |
| Rabbit | |
| 23 kDa | |
| 100 μL | |
| Cancer | |
| 3396 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q14197 | |
| ICT1 | |
| Synthetic peptides corresponding to ICT1(immature colon carcinoma transcript 1) The peptide sequence was selected from the middle region of ICT1. Peptide sequence AEWIAEPVRQKIAITHKNKINRLGELILTSESSRYQFRNLADCLQKIRDM. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction