missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICOS (CD278) Recombinant Rabbit Monoclonal Antibody (8P2G3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558130
This item is not returnable.
View return policy
Description
Immunogen sequence: EINGSADHRM FSFHNGGVQI SCKYPETVQQ LKMRLFRERE VLCELTKTKG SGNAVSIKNP MLCLYHLSNN SVSFFLNNPD SSQGSYYFCS LSIFDPPPFQ ERNLSGGYLH IYESQLCCQL KL.
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
| ICOS (CD278) | |
| Recombinant Monoclonal | |
| 1.63 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9R1T7, Q9WVS0 | |
| ICOS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-142 of mouse ICOS/CD278 (NP_059508.2). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8P2G3 | |
| Unconjugated | |
| ICOS | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54167, 64545 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction