Learn More
Scotsman™ Ice Flake Maker Ef103 Self Contained
Product Code. 12893216
Change view
Click to view available options
Capacity (Metric):
30 kg
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
12893216 30 kg Each
1 options
Product Code. 12893216 Supplier Scotsman™ Supplier No. EF103+EF103INSTALL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 108kg (238 lbs.) of flake ice production per day
  • Up to 30kg (66 lbs.) built-in ice storage
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • External components constructed from corrosion-resistant stainless steel crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-404A refrigerant
  • 120mm (4 12/16 in.) legs included, extensions available as option
  • Two years manufacturer warranty
  • Built -in PWD (pumped waste discharge) system for remote water drain (via easy-fit installation) for discharge of waste up to 1.5m above base or up to 20m from unit
Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Capacity (Metric) 30 kg